Learn More
Biolegend Ultra-LEAF™ Purified anti-HA.11 Epitope Tag
Supplier:  Biolegend 901521
Encompass_Preferred
Ultra-LEAF™ Purified anti-HA.11 Epitope Tag [16B12]; Isotype: Mouse IgG1, κ; Reactivity: YPYDVPDYA Tag; Apps: WB, Purification; Size: 100 ug
Catalog No. 50-207-5107