Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD200R
Supplier:  Biolegend 329315
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD200R [OX-108]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon; Apps: FC, Block; Size: 100 ug
Catalog No. 50-207-5862