Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD273 (B7-DC, PD-L2)
Supplier:  Biolegend 329623
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD273 (B7-DC, PD-L2) [24F.10C12]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IHC-F, FA; Size: 100 ug
Catalog No. 50-207-5888