Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD40
Supplier:  Biolegend 334350
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD40 [5C3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus, Squirrel Monkey ; Apps: FC, Costim; Size: 100 ug
Catalog No. 50-207-6176