Learn More
Biolegend Ultra-LEAF™ Purified anti-human C3a/C3a(desArg)/C3
Supplier:  Biolegend 518105
Encompass_Preferred
Ultra-LEAF™ Purified anti-human C3a/C3a(desArg)/C3 [K13/16]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, IP, WB, Neut; Size: 100 ug
Catalog No. 50-208-5908