Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD119 (IFN-γ R α chain)
Supplier:  Biolegend 308609
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD119 (IFN-γ R α chain) [GIR-208]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 ug
Catalog No. 50-207-3381