Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD11a
Supplier:  Biolegend 301233
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD11a [HI111]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Capuchin monkey, Chimpanzee, Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Sheep (Ovine); Apps: FC, IHC-F, WB, Block; Size: 100 ug
Catalog No. 50-207-2746