Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC, CyTOF, Block, IHC; Size: 1 mg
Supplier:  Biolegend 301632
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC, CyTOF, Block, IHC; Size: 1 mg
Catalog No. 50-237-6156