Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD16
Supplier:  Biolegend 302050
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Marmoset, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC, CyTOF, Block, IHC-F, IP, Stim; Size: 1 mg
Catalog No. 50-403-204