Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD178 (Fas-L)
Supplier:  Biolegend 306415
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD178 (Fas-L) [NOK-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon; Apps: FC, ICC, IP, WB; Size: 100 ug
Catalog No. 50-207-3284