Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD210 (IL-10 R)
Supplier:  Biolegend 308817
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD210 (IL-10 R) [3F9]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IP, Block; Size: 100 ug
Catalog No. 50-207-3388