Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 329716
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD274 (B7-H1, PD-L1) [29E.2A3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IHC, Block; Size: 1 mg
Catalog No. 50-403-329