Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD279 (PD-1)
Supplier:  Biolegend 329926
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD279 (PD-1) [EH12.2H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC, Block, IHC-F; Size: 1 mg
Catalog No. 50-403-332