Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD3
Supplier:  Biolegend 317325
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-403-286