Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD354 (TREM-1)
Supplier:  Biolegend 314914
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD354 (TREM-1) [TREM-26]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, Activ, WB; Size: 1 mg
Catalog No. 50-207-3696