Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD4
Supplier:  Biolegend 317453
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-207-3783