Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD4
Supplier:  Biolegend 300569
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD4 [RPA-T4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC, CyTOF, Activ, Block, IHC; Size: 100 ug
Catalog No. 50-207-2698