Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD49d
Supplier:  Biolegend 304339
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD49d [9F10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Common Marmoset, Cynomolgus, Dog (Canine), Horse (Equine), Rhesus, Sheep (Ovine), Squirrel Monkey; Apps: FC, IHC-F, Costim; Size: 100 ug
Catalog No. 50-207-3104