Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD56 (NCAM)
Supplier:  Biolegend 304631
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD56 (NCAM) [MEM-188]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cattle (Bovine, Cow); Apps: FC; Size: 100 ug
Catalog No. 50-207-3118