Learn More
Biolegend Ultra-LEAF™ Purified anti-human HLA-A,B,C
Supplier:  Biolegend 311427
Encompass_Preferred
Ultra-LEAF™ Purified anti-human HLA-A,B,C [W6/32]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Cynomolgus, Rhesus; Apps: FC, Activ, Block, IHC-F, IP, WB; Size: 100 ug
Catalog No. 50-207-3504