Learn More
Biolegend Ultra-LEAF™ Purified anti-human HLA-DR
Supplier:  Biolegend 307665
Encompass_Preferred
Ultra-LEAF™ Purified anti-human HLA-DR [L243]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Dog (Canine), Marmoset, Rhesus, Squirrel Monkey, Tamarin; Apps: FC, CyTOF, IP, WB, Block, IHC-F; Size: 100 ug
Catalog No. 50-207-3345