Learn More
Biolegend Ultra-LEAF™ Purified anti-human IFN-γ
Supplier:  Biolegend 507503
Encompass_Preferred
Ultra-LEAF™ Purified anti-human IFN-γ [MD-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ELISA Capture, ICFC, IHC-F, WB; Size: 100 ug
Catalog No. 50-208-5794