Learn More
Biolegend Ultra-LEAF™ Purified anti-human IL-12/IL-23 p40
Supplier:  Biolegend 508807
Encompass_Preferred
Ultra-LEAF™ Purified anti-human IL-12/IL-23 p40 [C8.6]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ELISA Capture, ICFC, IP, Neut, WB; Size: 100 ug
Catalog No. 50-208-5804