Learn More
Biolegend Ultra-LEAF™ Purified anti-human IL-16
Supplier:  Biolegend 519108
Encompass_Preferred
Ultra-LEAF™ Purified anti-human IL-16 [14.1]; Isotype: Mouse IgG2a; Reactivity: Human, Cross-Reactivity: Mouse; Apps: ICFC; Size: 1 mg
Catalog No. 50-208-5916