Learn More
Biolegend Ultra-LEAF™ Purified anti-human IL-4
Supplier:  Biolegend 500837
Encompass_Preferred
Ultra-LEAF™ Purified anti-human IL-4 [MP4-25D2]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine), Rhesus; Apps: ELISA, CyTOF, Neut, ICFC, IHC; Size: 100 ug
Catalog No. 50-208-5503