Learn More
Biolegend Ultra-LEAF™ Purified anti-human IL-6
Supplier:  Biolegend 501126
Encompass_Preferred
Ultra-LEAF™ Purified anti-human IL-6 [MQ2-13A5]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ELISA Capture, CyTOF, ELISPOT Capture, Neut, ICFC; Size: 1 mg
Catalog No. 50-208-5520