Learn More
Biolegend Ultra-LEAF™ Purified anti-human IL-7
Supplier:  Biolegend 501306
Encompass_Preferred
Ultra-LEAF™ Purified anti-human IL-7 [BVD10-40F6]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ELISA Capture, ELISPOT Capture, Neut, WB; Size: 1 mg
Catalog No. 50-208-5522