Learn More
Biolegend Ultra-LEAF™ Purified anti-human TNF-α
Supplier:  Biolegend 502805
Encompass_Preferred
Ultra-LEAF™ Purified anti-human TNF-α [MAb1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, ELISPOT Capture, Neut, WB; Size: 100 ug
Catalog No. 50-208-5575