Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD140b (PDGFRβ)
Supplier:  Biolegend 622803
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD140b (PDGFRβ) [A17169H]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: Block; Size: 100 ug
Catalog No. 50-208-6806