Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD279 (PD-1)
Supplier:  Biolegend 621604
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD279 (PD-1) [A17188B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: Block; Size: 1 mg
Catalog No. 50-208-6783