Learn More
Biolegend Ultra-LEAF™ Purified anti-human IL-13 [W20208A]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: Neut; Size: 100 ug
Supplier:  Biolegend 604153
Encompass_Preferred
Ultra-LEAF™ Purified anti-human IL-13 [W20208A]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: Neut; Size: 100 ug
Catalog No. 50-237-8322