Learn More
Biolegend Ultra-LEAF™ Purified anti-human/mouse Latent TGF-β
Supplier:  Biolegend 361203
Encompass_Preferred
Ultra-LEAF™ Purified anti-human/mouse Latent TGF-β [TW7-7H4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ELISA Capture; Size: 100 ug
Catalog No. 50-207-7988