Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD120b
Supplier:  Biolegend 358408
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD120b [3G7A02]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: Neut, FC; Size: 500 ug
Catalog No. 50-207-7751