Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD49b
Supplier:  Biolegend 103525
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD49b [HMα2]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F, IP, Block; Size: 100 ug
Catalog No. 50-206-9774