Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD80 [W17200C]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: Block, FC; Size: 100 ug
Supplier:  Biolegend 600053
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD80 [W17200C]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: Block, FC; Size: 100 ug
Catalog No. 50-237-7659