Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IL-2 Recombinant
Supplier:  Biolegend 937903
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IL-2 Recombinant [QA19A06]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: Neut; Size: 100 ug
Catalog No. 50-207-5380