Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD122 (IL-2Rβ)
Supplier:  Biolegend 123223
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD122 (IL-2Rβ) [TM-β1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IP, Block, Depletion; Size: 100 ug
Catalog No. 50-207-0499