Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD126 (IL-6Rα chain)
Supplier:  Biolegend 115816
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD126 (IL-6Rα chain) [D7715A7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, Block, IP, Neut; Size: 1 mg
Catalog No. 50-207-0210