Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD150 (SLAM)
Supplier:  Biolegend 115950
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD150 (SLAM) [TC15-12F12.2]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC, IP, Costim, Block; Size: 1 mg
Catalog No. 50-207-0223