Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD16/32
Supplier:  Biolegend 101330
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD16/32 [93]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC, IP, Block; Size: 1 mg
Catalog No. 50-603-882