Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC, IHC-F, IP, Block, Depletion; Size: 50 mg
Supplier:  Biolegend 102060
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC, IHC-F, IP, Block, Depletion; Size: 50 mg
Catalog No. 50-237-7479