Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD252 (OX40 Ligand)
Supplier:  Biolegend 108819
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD252 (OX40 Ligand) [RM134L]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-207-0065