Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD273 (B7-DC, PD-L2)
Supplier:  Biolegend 107221
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD273 (B7-DC, PD-L2) [TY25]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F, IP, WB; Size: 100 ug
Catalog No. 50-206-9978