Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD28
Supplier:  Biolegend 102121
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD28 [37.51]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC, IP, IHC-F, Costim, Block; Size: 5 mg
Catalog No. 50-206-9628