Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD3ε
Supplier:  Biolegend 100339
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F, IP, Activ, Block, WB, ICC; Size: 100 ug
Catalog No. 50-206-9447