Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD366 (Tim-3)
Supplier:  Biolegend 119731
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD366 (Tim-3) [RMT3-23]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, Block, IHC-F; Size: 100 ug
Catalog No. 50-207-0377