Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse/human IL-5
Supplier:  Biolegend 504315
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse/human IL-5 [TRFK5]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human, Cross-Reactivity: Guinea Pig; Apps: ELISA Capture, CyTOF, ELISPOT Capture, Neut, ICFC, IHC, WB; Size: 5 mg
Catalog No. 50-208-5644