Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse I-A/I-E
Supplier:  Biolegend 107655
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse I-A/I-E [M5/114.15.2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, IP, Block; Size: 100 ug
Catalog No. 50-207-0001