Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IFN-γ
Supplier:  Biolegend 517906
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IFN-γ [AN-18]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture; Size: 1 mg
Catalog No. 50-208-5907