Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse IFN-γ
Supplier:  Biolegend 513212
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse IFN-γ [H22]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: WB, Neut, ELISA, ICC, IP; Size: 100 mg
Catalog No. 50-208-5861